Tisk…

3-Nitro-L-phenylalanine 95%

Dodavatel: Thermo Scientific

Synonyms: (S)-2-Amino-3-(3-nitrophenyl)propanoic acid, H-Phe(3-NO₂)-OH

H63029.MD H63029.03 H63029.06
H63029.MDEA 1680 CZK
H63029.MD H63029.03 H63029.06
3-Nitro-L-phenylalanine 95%
3-Nitro-L-phenylalanine

Purity: 95% CAS No.: 19883-74-0 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white MDL No.: MFCD01860661

Vzorec: C₉H₁₀N₂O₄
MW: 210,189 g/mol
MDL Number: MFCD01860661
CAS číslo: 19883-74-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR