Tisk…

(S)-2-Aminopropanamide hydrochloride 95%

Dodavatel: Thermo Scientific

Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride

H63019.06 H63019.14 H63019.22
H63019.06EA 1820 CZK
H63019.06 H63019.14 H63019.22
(S)-2-Aminopropanamide hydrochloride 95%
(S)-2-Aminopropanamide hydrochloride

Purity: 95% CAS No.: 33208-99-0 Molecular Formula: C3H8N2O.HCl Formula Weight: 124.57 Physical Form: Solid Melting Point: 212-217° dec Optical Rotation: +11°(c=1 in methanol) MDL No.: MFCD00066145

Vzorec: C₃H₉ClN₂O
MW: 124,57 g/mol
Bod tání: 212…217 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00066145
CAS číslo: 33208-99-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR