Fmoc-O-(benzylphospho)-L-threonine 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
N-Fmoc-O-benzylphospho-L-threonine, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)(hydroxy)phosphoryl)oxy)butanoic acid, Fmoc-Thr(PO(OBzl)OH)-OH, Fmoc-Thr(HPO3Bzl)-OH
H63006.MD
H63006.03
H63006.MDEA
2830
CZK
H63006.MD
H63006.03
Fmoc-O-(benzylphospho)-L-threonine 97%
Fmoc-O-(benzylphospho)-L-threonine
Purity: 97%
CAS No.: 175291-56-2
Molecular Formula: C26H26NO8P
Formula Weight: 511.46
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Beilstein Reference: 7557104
MDL No.: MFCD00797870
Vzorec:
C₂₆H₂₆NO₈P MW: 511.467 g/mol |
MDL Number:
MFCD00797870 CAS číslo: 175291-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...