6-Methoxy-1,2,3,4-tetrahydroquinoline 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H63002.MD
H63002.03
H63002.06
H63002.MDEA
1240
CZK
H63002.MD
H63002.03
H63002.06
6-Methoxy-1,2,3,4-tetrahydroquinoline 97%
6-Methoxy-1,2,3,4-tetrahydroquinoline
Purity: 97%
CAS No.: 120-15-0
Molecular Formula: C10H13NO
Formula Weight: 163.22
Physical Form: Solid
Appearance: Yellow to brown
Melting Point: 34-37°
MDL No.: MFCD00090494
Vzorec:
C₁₀H₁₃NO Bod tání: 34…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00090494 CAS číslo: 120-15-0 EINECS: 204-374-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...