(R)-4-(1-aminoethyl)phenol 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H63001.03
H63001.06
H63001.03EA
6390
CZK
H63001.03
H63001.06
(R)-4-(1-aminoethyl)phenol 97%
(R)-4-(1-aminoethyl)phenol
Purity: 97%
CAS No.: 134855-89-3
Molecular Formula: C8H11NO
Formula Weight: 137.18
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
UN No.: UN3259
MDL No.: MFCD03844647
Vzorec:
C₈H₁₁NO |
MDL Number:
MFCD03844647 CAS číslo: 134855-89-3 ADR: 8,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...