Tisk…

Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Boc-Lys(Boc)-OH.DCHA, Boc-Lys(Boc)-OH·DCHA, Nα,Nepsilon-Di-Boc-L-lysine dicyclohexylammonium salt, Nɑ,N-ε-Di-Boc-L-lysine Dicyclohexylammonium Salt

H62785.14 H62785.22
H62785.14EA 2460 CZK
H62785.14 H62785.22
Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt 98%
Nalpha,N-epsilon-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt

Purity: 98% CAS No.: 15098-69-8 Molecular Formula: C16H30N2O6.C12H23N Formula Weight: 527.74 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: White Melting Point: 139-143° Beilstein Reference: 6262378 MDL No.: MFCD00038892

Vzorec: C₂₈H₅₃N₃O₆
Bod tání: 139…143 °C
MDL Number: MFCD00038892
CAS číslo: 15098-69-8
EINECS: 239-150-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR