Z-Dab(Boc)-OH·DCHA 98%
Dodavatel: Thermo Scientific
Synonyms:
(S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
H62772.03
H62772.06
H62772.03EA
3160
CZK
H62772.03
H62772.06
Z-Dab(Boc)-OH·DCHA 98%
Z-Dab(Boc)-OH·DCHA
Purity: 98%
CAS No.: 3350-13-8
Molecular Formula: C17H24N2O6.C12H23N
Formula Weight: 553.70
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00237341
Vzorec:
C₂₉H₄₇N₃O₆ MW: 533,71 g/mol |
MDL Number:
MFCD00237341 CAS číslo: 3350-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...