Methyl-3-bromo-2-fluorophenylacetate 96%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Methyl (3-bromo-2-fluorophenyl)acetate, 3-Bromo-2-fluorophenylacetic acid methyl ester
H62655.03
H62655.06
H62655.03EA
7180
CZK
H62655.03
H62655.06
Methyl-3-bromo-2-fluorophenylacetate 96%
Methyl-3-bromo-2-fluorophenylacetate
Purity: 96%
Molecular Formula: C9H8BrFO2
Formula Weight: 247.06
Physical Form: Solid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...