Tisk…

2-Amino-5-fluorobenzeneboronic acidpinacol ester 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Amino-5-fluorophenyl boronic acid pinacol ester, 4-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

H62575.MD H62575.03
H62575.MDEA 3750 CZK
H62575.MD H62575.03
2-Amino-5-fluorobenzeneboronic acidpinacol ester 96%
2-Amino-5-fluorobenzeneboronic acidpinacol ester

Purity: 96% CAS No.: 863578-24-9 Molecular Formula: C12H17BFNO2 Formula Weight: 237.08 Physical Form: Solid Appearance: Off-white Melting Point: 49-50° MDL No.: MFCD11617916

Vzorec: C₁₂H₁₇BFNO₂
MW: 237,081 g/mol
Bod tání: 49…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11617916
CAS číslo: 863578-24-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR