Tisk…

(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid 95%

Dodavatel: Thermo Scientific

Synonyms: Boc-D-Tle-OH, N-Boc-D-tert-leucine, Boc-D-Tle-O, (R)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid, (R)-2-(Boc-amino)-3,3-dimethylbutyric Acid, Boc-tBu-D-Gly-OH

H62473.03
H62473.03EA 4050 CZK
H62473.03
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid 95%
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid

Purity: 95% CAS No.: 124655-17-0 Molecular Formula: C11H21NO4 Formula Weight: 231.29 Physical Form: Solid MDL No.: MFCD00065575

Vzorec: C₁₁H₂₁NO₄
MW: 231,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065575
CAS číslo: 124655-17-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR