Boc-Asp-OtBu 95%
Dodavatel: Thermo Scientific
Synonyms:
Boc-Asp-OtBu, N-Boc-L-aspartic acid 1-tert-Butyl ester, N-(tert-Butoxycarbonyl)-L-aspartic acid 1-tert-Butyl ester, 1-tert-Butyl N-Boc-L-aspartate, 1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate, N-Boc-L-aspartic acid 1-tert-butyl ester
Purity: 95%
CAS No.: 34582-32-6
Molecular Formula: C13H23NO6
Formula Weight: 289.32
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Odour: Odourless
Beilstein Reference: 4191701
Optical Rotation: -24°(c=1 in methanol)
MDL No.: MFCD00038272
Vzorec:
C₁₃H₂₃NO₆ MW: 289,328 g/mol |
MDL Number:
MFCD00038272 CAS číslo: 34582-32-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...