Tisk…

(S)-2-Amino-3-phenylpropanamide 98%

Dodavatel: Thermo Scientific

Synonyms: H-Phe-NH₂, L-Phenylalaninamide

H62030.03 H62030.06
H62030.03EA 2020 CZK
H62030.03 H62030.06
(S)-2-Amino-3-phenylpropanamide 98%
(S)-2-Amino-3-phenylpropanamide

Purity: 98% CAS No.: 5241-58-7 Molecular Formula: C9H12N2O Formula Weight: 164.20 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Beilstein Reference: 6270941 MDL No.: MFCD00038146

Vzorec: C₉H₁₂N₂O
MW: 164,20 g/mol
MDL Number: MFCD00038146
CAS číslo: 5241-58-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR