(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%
Dodavatel: Thermo Scientific
Synonyms:
Boc-Lys(Boc)-OH, Nα,Nepsilon-Di-Boc-L-lysine
H62019.22
H62019.14
H62019.22EA
7860
CZK
H62019.22
H62019.14
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid 98%
(S)-2,6-Bis((tert-butoxycarbonyl)amino)hexanoic acid
Purity: 98%
CAS No.: 2483-46-7
Molecular Formula: C16H30N2O6
Formula Weight: 346.42
Storage Temperature: -30°C to -10°C
Physical Form: Low melting solid
Appearance: White
MDL No.: MFCD00038515
Vzorec:
C₁₆H₃₀N₂O₆ MW: 346,4 g/mol |
MDL Number:
MFCD00038515 CAS číslo: 2483-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...