4-Amino-2,6-dimethylpyridine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2,6-Dimethylpyridin-4-amine
H61970.03
H61970.06
H61970.03EA
3710
CZK
H61970.03
H61970.06
4-Amino-2,6-dimethylpyridine 98%
4-Amino-2,6-dimethylpyridine
Purity: 98%
CAS No.: 3512-80-9
Molecular Formula: C7H10N2
Formula Weight: 122.17
Physical Form: Solid
UN No.: UN2811
MDL No.: MFCD00130078
Vzorec:
C₇H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00130078 CAS číslo: 3512-80-9 EINECS: 225-515-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...