Tisk…
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 98%
Dodavatel: Thermo Fisher Scientific
Synonyms: Boc-Tyr(Me)-OH, N-Boc-O-methyl-L-tyrosine
H61820.03 H61820.06
H61820.03EA 2110 CZK
H61820.03 H61820.06
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-methoxyphenyl)propanoic acid
Purity: 98% CAS No.: 53267-93-9 Molecular Formula: C15H21NO5 Formula Weight: 295.33 Physical Form: Solid MDL No.: MFCD00065603
Vzorec: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065603
CAS číslo: 53267-93-9