4,6-Dichloro-5-fluoropyrimidine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H61770.03
H61770.06
H61770.03EA
6040
CZK
H61770.03
H61770.06
4,6-Dichloro-5-fluoropyrimidine 98%
4,6-Dichloro-5-fluoropyrimidine
Purity: 98%
CAS No.: 213265-83-9
Molecular Formula: C4HCl2FN2
Formula Weight: 166.97
Flash Point: 91°(196°F)
Physical Form: Liquid
Boiling Point: 50-60°/14mm
UN No.: UN3265
MDL No.: MFCD08056331
Vzorec:
C₄HCl₂FN₂ MW: 166,97 g/mol Bod varu: 50…60 °C (14 mmHg) Bod vzplanutí: 91 °C (196 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08056331 CAS číslo: 213265-83-9 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...