Boc-Lys(Fmoc)-OH 98%
Dodavatel: Thermo Scientific
Synonyms:
N-epsilon-FMOC-N-alpha-BOC-L-Lysine
H61126.06
H61126.14
H61126.06EA
3190
CZK
H61126.06
H61126.14
Boc-Lys(Fmoc)-OH 98%
Boc-Lys(Fmoc)-OH
Purity: 98%
CAS No.: 84624-27-1
Molecular Formula: C26H32N2O6
Formula Weight: 468.55
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White
Odour: Odourless
Melting Point: 93-98°
Beilstein Reference: 3576584
MDL No.: MFCD00062032
Vzorec:
C₂₆H₃₂N₂O₆ MW: 468,55 g/mol Bod tání: 93…98 °C |
MDL Number:
MFCD00062032 CAS číslo: 84624-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...