Ytterbium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Packaged under argon.
45920.04
45920.09
45920.18
45920.04EA
5630
CZK
45920.04
45920.09
45920.18
Ytterbium ≥99.9% (REO, rare earth oxide basis), powder < 200 mesh
Ytterbium
Purity: 99.9%
CAS No.: 7440-64-4
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00011286
Vzorec:
Yb MW: 173.04 Bod varu: 1427 Bod tání: 824 Storage Temperature: Ambient |
MDL Number:
MFCD00011286 CAS číslo: 7440-64-4 EINECS: 231-173-2 UN: 3089 ADR: 4.1 II Merck Index: 12,10239 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...