N-Boc-4-bromo-3-fluoroaniline 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
tert-Butyl N-(4-bromo-3-fluorophenyl)carbamate, tert-Butyl (4-bromo-3-fluorophenyl)carbamate
H59398.03
H59398.03EA
9580
CZK
H59398.03
N-Boc-4-bromo-3-fluoroaniline 96%
N-Boc-4-bromo-3-fluoroaniline
Purity: 96%
CAS No.: 868735-43-7
Molecular Formula: C11H13BrFNO2
Formula Weight: 290.13
Melting Point: 68-70°
Beilstein Reference: 14460051
UN No.: UN2811
MDL No.: MFCD11976284
Vzorec:
C₁₁H₁₃BrFNO₂ MW: 290,132 g/mol Bod tání: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD11976284 CAS číslo: 868735-43-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...