3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester 97%
Dodavatel: Thermo Fisher Scientific
H58731.03
H58731.06
H58731.03EA
2200
CZK
H58731.03
H58731.06
3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester 97%
3-[2-(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 915402-03-8
Molecular Formula: C17H25BO2Si
Formula Weight: 300.28
Melting Point: 86-90°
MDL No.: MFCD08706285
Vzorec:
C₁₇H₂₅BO₂Si Bod tání: 86…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD08706285 CAS číslo: 915402-03-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...