N-(Benzoyloxy)succinimide 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,5-Dioxopyrrolidin-1-yl benzoate, Bz-OSu
H58625.03
H58625.06
H58625.03EA
2700
CZK
H58625.03
H58625.06
N-(Benzoyloxy)succinimide 97%
N-(Benzoyloxy)succinimide
Purity: 97%
CAS No.: 23405-15-4
Molecular Formula: C11H9NO4
Formula Weight: 219.19
Storage Temperature: 2°C to 8°C
MDL No.: MFCD00078953
Vzorec:
C₁₁H₉NO₄ MW: 219,196 g/mol |
MDL Number:
MFCD00078953 CAS číslo: 23405-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...