Tisk…

4-(Azidomethyl)benzeneboronic acid pinacol ester 95%

Dodavatel: Thermo Fisher Scientific

H58401.03 H58401.06
H58401.03EA 3130 CZK
H58401.03 H58401.06
4-(Azidomethyl)benzeneboronic acid pinacol ester 95%
4-(Azidomethyl)benzeneboronic acid pinacol ester

Purity: 95% CAS No.: 1239481-05-0 Molecular Formula: C13H19BN3O2 Formula Weight: 260.12 MDL No.: MFCD20922785

Vzorec: C₁₃H₁₉BN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD20922785
CAS číslo: 1239481-05-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR