(S)-(+)-Pyrrolidine-3-carboxylic acid ≥98%
Dodavatel: Thermo Fisher Scientific
H57895.MD
H57895.03
H57895.MDEA
2660
CZK
H57895.MD
H57895.03
(S)-(+)-Pyrrolidine-3-carboxylic acid ≥98%
(S)-(+)-Pyrrolidine-3-carboxylic acid
Purity: 98+%
CAS No.: 72580-53-1
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Storage Temperature: 2°C to 8°C
Melting Point: 185-191°
Beilstein Reference: 3537293
Optical Rotation: +18° (c=2 in water)
MDL No.: MFCD06738463
Vzorec:
C₅H₉NO₂ Bod tání: 185…191 °C |
MDL Number:
MFCD06738463 CAS číslo: 72580-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...