Tisk…

1-Benzyl-5-bromoindole 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Benzyl-5-bromo-1H-indole

H57676.03 H57676.06
H57676.03EA 2650 CZK
H57676.03 H57676.06
1-Benzyl-5-bromoindole 97%
1-Benzyl-5-bromoindole

Purity: 97% CAS No.: 10075-51-1 Molecular Formula: C15H12BrN Formula Weight: 286.17 MDL No.: MFCD04337704

Vzorec: C₁₅H₁₂BrN
MW: 286,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04337704
CAS číslo: 10075-51-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR