2-Oxa-6-azaspiro[3.4]octane 95%
Dodavatel: Thermo Scientific
|
Varování
|
H57579.MD
H57579.ME
H57579.MDEA
9410
CZK
H57579.MD
H57579.ME
2-Oxa-6-azaspiro[3.4]octane 95%
2-Oxa-6-azaspiro[3.4]octane
Purity: 95%
CAS No.: 220290-68-6
Molecular Formula: C6H11NO
Formula Weight: 113.16
Flash Point: 63°(145°F)
Boiling Point: 187°
MDL No.: MFCD14586455
Vzorec:
C₆H₁₁NO Bod varu: 187 °C Hustota: 1,08 Bod vzplanutí: 63 °C (145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD14586455 CAS číslo: 220290-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...