Tisk…

Methyl-2-oxoindane-1-carboxylate 97%

Dodavatel: Thermo Fisher Scientific
Varování

H57535.03 H57535.06
H57535.03EA 2080 CZK
H57535.03 H57535.06
Methyl-2-oxoindane-1-carboxylate 97%
Methyl-2-oxoindane-1-carboxylate

Purity: 97% CAS No.: 104620-34-0 Molecular Formula: C11H10O3 Formula Weight: 190.20 Storage Temperature: 2°C to 8°C Melting Point: 63-70° MDL No.: MFCD06200715

Vzorec: C₁₁H₁₀O₃
Bod tání: 63…70 °C
MDL Number: MFCD06200715
CAS číslo: 104620-34-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR