Tisk…

2-n-Propylheptanoic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

H57414.MD H57414.03
H57414.MDEA 1950 CZK
H57414.MD H57414.03
2-n-Propylheptanoic acid 97%
2-n-Propylheptanoic acid

Purity: 97% CAS No.: 31080-39-4 Molecular Formula: C10H20O2 Formula Weight: 172.27 Physical Form: Liquid Appearance: Colourless to pale yellow MDL No.: MFCD01723758

Vzorec: C₁₀H₂₀O₂
MW: 172.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01723758
CAS číslo: 31080-39-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR