Tisk…

[3-(Bromomethyl)oxetan-3-yl]methanol 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: (3-(Bromomethyl)oxetan-3-yl)methanol

H57413.MD H57413.ME
H57413.MDEA 1710 CZK
H57413.MD H57413.ME
[3-(Bromomethyl)oxetan-3-yl]methanol 95%
[3-(Bromomethyl)oxetan-3-yl]methanol

Purity: 95% CAS No.: 22633-44-9 Molecular Formula: C5H9BrO2 Formula Weight: 181.03 Flash Point: 100°(212°F) Boiling Point: 242° MDL No.: MFCD09800598

Vzorec: C₅H₉BrO₂
MW: 181,04 g/mol
Bod varu: 242 °C
Hustota: 1,598
Bod vzplanutí: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD09800598
CAS číslo: 22633-44-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR