Tisk…

Pyrrolidine-3-carboxylic acid hydrochloride 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H57365.MD H57365.03
H57365.MDEA 930 CZK
H57365.MD H57365.03
Pyrrolidine-3-carboxylic acid hydrochloride 95%
Pyrrolidine-3-carboxylic acid hydrochloride

Purity: 95% CAS No.: 953079-94-2 Molecular Formula: C5H9NO2.HCl Formula Weight: 151.59 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 30-38° MDL No.: MFCD06801272

Vzorec: C₅H₁₀ClNO₂
Bod tání: 30…38 °C
Bod vzplanutí: >110 °C (230 °F)
MDL Number: MFCD06801272
CAS číslo: 953079-94-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR