D-β-Proline ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
(R)-Pyrrolidine-3-carboxylic acid, H-D-β-Pro-OH
H57356.MD
H57356.03
H57356.MDEA
3570
CZK
H57356.MD
H57356.03
D-β-Proline ≥98%
D-beta-Proline
Purity: 98+%
CAS No.: 72580-54-2
Molecular Formula: C5H9NO2
Formula Weight: 115.13
Storage Temperature: 2°C to 8°C
Melting Point: 185-191°
Beilstein Reference: 5496144
Optical Rotation: -18° (c=2 in water)
MDL No.: MFCD08272856
Vzorec:
C₅H₉NO₂ MW: 115.13 g/mol Bod tání: 185…191 °C |
MDL Number:
MFCD08272856 CAS číslo: 72580-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...