Tisk…

(S)-Pyrrolidin-3-ylmethanol 95%

Dodavatel: Thermo Scientific
Varování

H57199.MD H57199.03
H57199.MDEA 3400 CZK
H57199.MD H57199.03
(S)-Pyrrolidin-3-ylmethanol 95%
(S)-Pyrrolidin-3-ylmethanol

Purity: 95% CAS No.: 110013-19-9 Molecular Formula: C5H11NO Formula Weight: 101.15 Storage Temperature: 2°C to 8°C Melting Point: 174-176° MDL No.: MFCD09260722

Vzorec: C₅H₁₁NO
MW: 101,15 g/mol
Bod tání: 174…176 °C
Hustota: 0,979
MDL Number: MFCD09260722
CAS číslo: 110013-19-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR