2,7-Dibromo-9,9-dihexylfluorene 98%
Dodavatel: Thermo Scientific
Synonyms:
2,7-Dibromo-9,9-dihexyl-9H-fluorene
H56952.14
H56952.22
H56952.14EA
8210
CZK
H56952.14
H56952.22
2,7-Dibromo-9,9-dihexylfluorene 98%
2,7-Dibromo-9,9-dihexylfluorene
Purity: 98%
CAS No.: 189367-54-2
Molecular Formula: C25H32Br2
Formula Weight: 492.33
Melting Point: 67-71°
MDL No.: MFCD03427215
Vzorec:
C₂₅H₃₂Br₂ MW: 492,34 g/mol Bod tání: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427215 CAS číslo: 189367-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...