Sodium phenylpyruvate 98%
Dodavatel: Thermo Scientific
Synonyms:
2-Oxo-3-phenylpropionic acid sodium salt, Phenylpyruvic acid sodium salt
H56767.06
H56767.14
H56767.06EA
2060
CZK
H56767.06
H56767.14
Sodium phenylpyruvate 98%
Sodium phenylpyruvate
Phenylpyruvic acid, sodium salt is a substrate for phenylpyruvate decarboxylase and phenylpyruvate tautomerase.
Purity: 98%
CAS No.: 114-76-1
Molecular Formula: C9H7NaO3
Formula Weight: 186.14
Storage Temperature: 2°C to 8°C
Melting Point:
Vzorec:
C₉H₇NaO₃ MW: 186,14 g/mol Bod tání: >300 °C |
MDL Number:
MFCD00002590 CAS číslo: 114-76-1 EINECS: 204-053-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...