Tisk…

3-Boc-3-methylamino azetidine hydrochloride 95%

Dodavatel: Thermo Scientific
Varování

H56060.MD H56060.03
H56060.MDEA 2770 CZK
H56060.MD H56060.03
3-Boc-3-methylamino azetidine hydrochloride 95%
3-Boc-3-methylamino azetidine hydrochloride

Purity: 95% CAS No.: 577777-20-9 Molecular Formula: C9H18N2O2.HCl Formula Weight: 222.72 MDL No.: MFCD12963843

Vzorec: C₉H₁₉ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12963843
CAS číslo: 577777-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR