1-Cbz-piperazine 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H56008.AA
H56008.AC
H56008.AE
H56008.AAEA
2180
CZK
H56008.AA
H56008.AC
H56008.AE
1-Cbz-piperazine 98%
1-Cbz-piperazine
Purity: 98%
CAS No.: 31166-44-6
Molecular Formula: C12H16N2O2
Formula Weight: 220.27
Refractive Index: 1.5460
Boiling Point: 158-161°/1.4mm
Beilstein Reference: 179500
MDL No.: MFCD00274317
Vzorec:
C₁₂H₁₆N₂O₂ Bod varu: 158…161 °C (1,4 mmHg) Hustota: 1,142 Storage Temperature: Ambient |
MDL Number:
MFCD00274317 CAS číslo: 31166-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...