Tisk…

2-(1-Adamantyl)-4-methylphenol 99%

Dodavatel: Thermo Scientific
Varování

H55922.03 H55922.06
H55922.03EA 2940 CZK
H55922.03 H55922.06
2-(1-Adamantyl)-4-methylphenol 99%
2-(1-Adamantyl)-4-methylphenol

Purity: 99% CAS No.: 41031-50-9 Molecular Formula: C17H22O Formula Weight: 242.36 Physical Form: Powder Appearance: White Melting Point: 128-130° MDL No.: MFCD00168147

Vzorec: C₁₇H₂₂O
MW: 242,362 g/mol
Bod tání: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00168147
CAS číslo: 41031-50-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR