Tisk…

3-Amino-1-propanesulphonic acid 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Tramiprosate, Homotaurine, 3-Aminopropane-1-sulphonic acid, 3APS

H55918.03 H55918.06 H55918.14
H55918.03EA 1440 CZK
H55918.03 H55918.06 H55918.14
3-Amino-1-propanesulphonic acid 97%
3-Amino-1-propanesulphonic acid

Purity: 97% CAS No.: 3687-18-1 Molecular Formula: C3H9NO3S Formula Weight: 139.17 Melting Point: 293° dec MDL No.: MFCD00008225

Vzorec: C₃H₉NO₃S
MW: 139,17 g/mol
Bod tání: 293 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00008225
CAS číslo: 3687-18-1
EINECS: 222-977-4
Merck Index: 14,10218

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR