Tisk…

1-(1-Naphthylmethyl)piperazine 97%

Dodavatel: Thermo Fisher Scientific
Varování

H55398.03 H55398.06 H55398.14
H55398.03EA 1940 CZK
H55398.03 H55398.06 H55398.14
1-(1-Naphthylmethyl)piperazine 97%
1-(1-Naphthylmethyl)piperazine

Purity: 97% CAS No.: 40675-81-8 Molecular Formula: C15H18N2 Formula Weight: 226.32 Physical Form: Powder Appearance: White to brown Melting Point: 63-67° UN No.: UN3077 MDL No.: MFCD01314185

Vzorec: C₁₅H₁₈N₂
Bod tání: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01314185
CAS číslo: 40675-81-8
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR