Tisk…

Neodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh

Dodavatel: Thermo Scientific
Nebezpečí


Packaged under argon.

45914.14 45914.06
45914.14EA 20350 CZK
45914.14 45914.06
Neodymium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Neodymium

Purity: 99.9% CAS No.: 7440-00-8 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Grey to silver Odour: Odourless UN No.: 1383 MDL No.: MFCD00011130

Vzorec: Nd
MW: 144,24 g/mol
Bod varu: 2474 °C (1013 hPa)
Bod tání: 1014,85 °C
Hustota: 6,46 g/cm³ (1240 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS číslo: 7440-00-8
EINECS: 231-109-3
UN: 1383
ADR: 4.2,I
Merck Index: 12,06538

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR