tert-Butyl-4-oxoazepane-1-carboxylate 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tert-butyl 4-oxoazepane-1-carboxylate, 4-Oxoazepane-1-carboxylic acid tert-butyl ester, N-BOC-Hexahydro-1H-azepin-4-one, tert-Butyl 4-oxoazepane-1-carboxylate
H55214.03
H55214.06
H55214.03EA
3180
CZK
H55214.03
H55214.06
tert-Butyl-4-oxoazepane-1-carboxylate 98%
tert-Butyl-4-oxoazepane-1-carboxylate
Purity: 98%
CAS No.: 188975-88-4
Molecular Formula: C11H19NO3
Formula Weight: 213.28
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4670
MDL No.: MFCD03788435
Vzorec:
C₁₁H₁₉NO₃ MW: 213,277 g/mol |
MDL Number:
MFCD03788435 CAS číslo: 188975-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...