tert-Butyl (2-bromoethyl)carbamate 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-(BOC-amino)ethyl bromide
H55211.03
H55211.06
H55211.03EA
2240
CZK
H55211.03
H55211.06
tert-Butyl (2-bromoethyl)carbamate 96%
tert-Butyl (2-bromoethyl)carbamate
Purity: 96%
CAS No.: 39684-80-5
Molecular Formula: C7H14BrNO2
Formula Weight: 224.10
Storage Temperature: -30°C to -10°C
Melting Point: 30-32°
Beilstein Reference: 2325117
MDL No.: MFCD02683428
Vzorec:
C₇H₁₄BrNO₂ MW: 224,10 g/mol Bod tání: 30…32 °C |
MDL Number:
MFCD02683428 CAS číslo: 39684-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...