Bis(4-tert-butylphenyl)iodonium-p-toluenesulphonate 99+% (metals basis), Electronic Grade
Dodavatel: Thermo Fisher Scientific
H55188.03
H55188.06
H55188.03EA
4050
CZK
H55188.03
H55188.06
Bis(4-tert-butylphenyl)iodonium-p-toluenesulphonate 99+% (metals basis), Electronic Grade
Bis(4-tert-butylphenyl)iodonium-p-toluenesulphonate
Purity: 99+%
CAS No.: 131717-99-2
Molecular Formula: C27H33IO3S
Formula Weight: 564.52
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: 237-240°
MDL No.: MFCD02683472
Vzorec:
C₂₇H₃₃IO₃S MW: 564.522 g/mol Bod tání: 237…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683472 CAS číslo: 131717-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...