Tisk…

Silver(I) acetylacetonate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Silver 2,4-pentanedionate, Silver acetylacetonate

H55131.03 H55131.06 H55131.14
H55131.03EA 2650 CZK
H55131.03 H55131.06 H55131.14
Silver(I) acetylacetonate 98%
Silver(I) acetylacetonate

Purity: 98% CAS No.: 15525-64-1 Molecular Formula: C5H7AgO2 Formula Weight: 206.98 Storage Temperature: 2°C to 8°C Odour: Characteristic Melting Point: 100° dec. MDL No.: MFCD00000012

Vzorec: C₅H₈AgO₂
MW: 207,985 g/mol
Bod tání: 100 °C (rozkl.)
MDL Number: MFCD00000012
CAS číslo: 15525-64-1
EINECS: 239-568-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR