Pyrimidine-2-carbothioamide 97%
Dodavatel: Thermo Scientific
|
Varování
|
H54783.03
H54783.06
H54783.03EA
2640
CZK
H54783.03
H54783.06
Pyrimidine-2-carbothioamide 97%
Pyrimidine-2-carbothioamide
Purity: 97%
CAS No.: 4537-73-9
Molecular Formula: C5H5N3S
Formula Weight: 139.17
Beilstein Reference: 508162
MDL No.: MFCD12093703
Vzorec:
C₅H₅N₃S Storage Temperature: Ambient |
MDL Number:
MFCD12093703 CAS číslo: 4537-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...