Tisk…

2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)picolinamide

H54614.MD H54614.03
H54614.MDEA 3020 CZK
H54614.MD H54614.03
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1006876-27-2 Molecular Formula: C14H21BN2O3 Formula Weight: 276.14 MDL No.: MFCD08458484

Vzorec: C₁₄H₂₁BN₂O₃
MW: 276.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458484
CAS číslo: 1006876-27-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR