2-(3-Pyridyl)benzimidazole-6-carboxylic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
H54593.MD
H54593.03
H54593.06
H54593.MDEA
2390
CZK
H54593.MD
H54593.03
H54593.06
2-(3-Pyridyl)benzimidazole-6-carboxylic acid 97%
2-(3-Pyridyl)benzimidazole-6-carboxylic acid
Purity: 97%
CAS No.: 312496-16-5
Molecular Formula: C13H9N3O2
Formula Weight: 239.23
MDL No.: MFCD01414693
Vzorec:
C₁₃H₉N₃O₂ MW: 239,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01414693 CAS číslo: 312496-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...