Tisk…

Pyrimidine-5-carboxaldehyde 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 5-Pyrimidinecarboxaldehyde

H54448.03 H54448.06 H54448.14
H54448.03EA 3260 CZK
H54448.03 H54448.06 H54448.14
Pyrimidine-5-carboxaldehyde 97%
Pyrimidine-5-carboxaldehyde

Purity: 97% CAS No.: 10070-92-5 Molecular Formula: C5H4N2O Formula Weight: 108.10 Melting Point: 39-43° Boiling Point: 80-82°/4mm MDL No.: MFCD03426065

Vzorec: C₅H₄N₂O
Bod varu: 80…82 °C (4 mmHg)
Bod tání: 39…43 °C
Hustota: 1,23
Storage Temperature: Refrigerator
MDL Number: MFCD03426065
CAS číslo: 10070-92-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR