Tisk…

2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%

Dodavatel: Thermo Fisher Scientific
Varování

H54151.MD H54151.03
H54151.MDEA 1830 CZK
H54151.MD H54151.03
2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester 96%
2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester

Purity: 96% CAS No.: 1006876-28-3 Molecular Formula: C14H21BN2O3 Formula Weight: 276.14 Storage Temperature: 2°C to 8°C Melting Point: 52-54° MDL No.: MFCD13191367

Vzorec: C₁₄H₂₁BN₂O₃
Bod tání: 52…54 °C
MDL Number: MFCD13191367
CAS číslo: 1006876-28-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR