3-(4-tert-Butylphenyl)propionic acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(4-(tert-Butyl)phenyl)propanoic acid
H54011.03
H54011.06
H54011.14
H54011.03EA
1650
CZK
H54011.03
H54011.06
H54011.14
3-(4-tert-Butylphenyl)propionic acid 97%
3-(4-tert-Butylphenyl)propionic acid
Purity: 97%
CAS No.: 1208-64-6
Molecular Formula: C13H18O2
Formula Weight: 206.28
Melting Point: 102°
MDL No.: MFCD00796482
Vzorec:
C₁₃H₁₈O₂ MW: 206,29 g/mol Bod tání: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00796482 CAS číslo: 1208-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...