4-Diethylamino-2-butyn-1-ol 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H53501.06
H53501.14
H53501.06EA
1810
CZK
H53501.06
H53501.14
4-Diethylamino-2-butyn-1-ol 98%
4-Diethylamino-2-butyn-1-ol
Purity: 98%
CAS No.: 10575-25-4
Molecular Formula: C8H15NO
Formula Weight: 141.21
Flash Point: 104°(219°F)
Refractive Index: 1.479
Boiling Point: 231-232°
MDL No.: MFCD00671356
Vzorec:
C₈H₁₅NO Bod varu: 231…232 °C Hustota: 0,952 Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00671356 CAS číslo: 10575-25-4 EINECS: 234-163-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...